Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ MAS™ UA Controls
Bi-level control designed to monitor urine dipstick and microscopic components
Includes: Includes 10 commonly tested dipstick tests plus microalbumin, creatinine and hCG
  • Products 5
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  
23-038-380
Available on GSA/VA Contract for Federal Government customers only.  SDS
Thermo Scientific
UAB115
Assayed; Level 1; Normal; 6 x 15mL
N/A N/A N/A  
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 
23-038-381
Available on GSA/VA Contract for Federal Government customers only.  SDS
Thermo Scientific
UAB215
6 x 15mL
N/A N/A N/A  
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 
23-038-384
Available on GSA/VA Contract for Federal Government customers only.  SDS
Thermo Scientific
UAB-160
Assayed; Level 1; Normal; 4 x 60mL
N/A N/A N/A  
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 
23-038-385
Available on GSA/VA Contract for Federal Government customers only.  SDS
Thermo Scientific
UAB-260
4 x 60mL
N/A N/A N/A  
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 
23-038-383
Available on GSA/VA Contract for Federal Government customers only.  SDS
Thermo Scientific
UABMP
Assayed; Bi-level; Multi-Pack; 6 x 15mL
N/A N/A N/A  
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.  This product is regulated by the federal government, and may not be ordered online. 
Descriptions & Specifications
Related Products