Learn More
Thermo Scientific™ MaxQ™ 2506 Reciprocating Shaker Package
Flexible, economical analog units are ideal for chemical extractions, mixing blood samples, and silver staining of polyacrylamide gels
Manufacturer:  Thermo Scientific™ SHKA2506PROMO
Catalog No. 11-676-064
Add to cart
Alert:
Products on this page may include voltages not considered standard in North America. Please review carefully.
Description
Description
Units feature an easy-to-use analog operating system and variable speed control for gentle to vigorous agitation. Built-in timer ensures repeatable results. Three interchangeable platforms provide increased capacity.
  • Back-and-forth motion with adjustable stroke length (0.5 to 1.5 in. [1.2 to 3.8cm]) automates hand shaking of separatory funnels
  • Can be used in incubators, warm rooms, environmental chambers and refrigerators
  • Operates in continuous or timed (1 to 60 min.) mode
  • Metal locking tab on speed control knob prevents speed from increasing or decreasing if knob is bumped
  • DC motor and variable speed (10 to 250rpm) controlled by rotary dial
Promotional Package:
With Platform and Clamp Starter Kit
Applications:
  • Chemical extractions
  • Mixing blood samples
  • Silver staining of polyacrylamide gels
Certifications
UL, cUL
Warranty and Services
24 months
Specifications
Specifications
MaxQ 2506
1.2 to 3.8cm
39 to 104°F
1 to 60 min.
27 lb.
DC
UL, cUL
Ideal for chemical extractions, mixing blood samples and Silver staining of Polyacrylamide gels
1.2 to 3.8cm
120V 50/60Hz
7.2 in.
17.4 in.
13.5 in.
10 to 250rpm
120V
4 to 40°C
20 to 80%
12.3kg
50/60Hz
Analog
0.5 to 1.5 in.
6.8kg
17.4 x 13.5 x 7.2 in. (44.2 x 34.3 x 18.3cm)
18.3cm
44.2cm
34.3cm