Learn More
Thermo Scientific™ MaxQ™ 2508 Dual-Action Orbital Reciprocating Shaker Package
Affordable analog unit with back-and-forth or circular motion for a wide range of shaking applications
Manufacturer:  Thermo Scientific™ SHKA2508PROMO
Catalog No. 11-676-065
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
Products on this page may include voltages not considered standard in North America. Please review carefully.
Description
Description
Variable speed control allows gentle to vigorous agitation. Built-in timer facilitates repeatable results. The versatile back-and-forth or circular orbital motion is ideal for a wide range of applications. A control lever changes the motion while the shaker is stationary or in operation.
  • Shaking motion: back-and-forth, with fixed stroke angle of 0.4 in. (1.2cm) for shaking of separatory funnels, or fixed circular motion of 0.4 in. (1.2cm) for growing bacteria and yeast
  • Can be used in incubators, warm rooms, environmental chambers and refrigerators
  • Operates in temperature ranges of 4° to 40°C and humidity conditions of 20 to 80% noncondensing
  • Option between Reciprocating or Orbital and Timed (1 to 60 min.) or continuous
  • Metal locking tab on speed control knob prevents increased or decreased speed if the knob is bumped
  • DC motor and variable speed (40 to 400rpm) controlled by rotary dial
Promotional Package:
With Platform and Clamp Starter Kit
Applications:
Growing bacteria, Dissolution studies, Staining of electrophoresis gels, Chemical extractions
Certifications
UL, cUL, CE
Warranty and Services
24 months
Specifications
Specifications
MaxQ 2508
1.2cm
39 to 104°F
1 to 60 min.
38 lb.
DC
UL, cUL, CE
Ideal for growing bacteria, dissolution studies and staining of electrophoresis gels and chemical extractions
1.2cm
120V 50/60Hz
7 in.
17.4 in.
13.5 in.
40 to 400rpm
120V
4 to 40°C
20 to 80% Non-condensing
17.2kg
50/60Hz
Analog
0.4 in.
6.8kg
17.4 x 13.5 x 7 in. (44.2 x 34.3 x 17.8cm)
17.8cm
44.2cm
34.3cm