Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Thermo Scientific™ MaxQ™ 4000 Benchtop Orbital Shaker Packages
Ideal combination of incubated/refrigerated temperatures with orbital shaking
Manufacturer:  Thermo Scientific™ SHKA4000PROMO
 View more versions of this product
Catalog No. 11-676-073
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
Products on this page may include voltages not considered standard in North America. Please review carefully.
Requires : Universal Platform (without clamps) or Dedicated Platform (with pre-installed clamps). See attached brochure for more details.
Specifications Description & Specifications
Specifications
SHKA4000
1.9cm
18 in.
45.72cm
53.3cm
57.2cm
50/60Hz
167 lb.
1 to 60 min.
32 in.
50.8cm
60.9cm
13 in.
20 in.
±0.1°C
Flask 2L
120V 50/60Hz
40 to 400rpm
18 in.
45.72cm
21 in.
22.5 in.
33cm
50 lb.
75.8kg
120V
81.3cm
24 in.
LED
Benchtop incubated shaker
CE, cUL, UL
Analog
Platform and clamp starter kit
10° above ambient to 60°C