Learn More
Thermo Scientific™ MaxQ™ HP Tabletop Orbital Shaker Packages
Operates in reach-in incubators, environmental rooms, or on lab benches
Manufacturer:  Thermo Scientific™ SHKE416HPPR
 View more versions of this product
Catalog No. 11-676-238
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Alert:
Products on this page may include voltages not considered standard in North America. Please review carefully.
Description
Description
MaxQ HP Tabletop orbital shakers combine high durability and capacity with unequalled ease of use. The proprietary triple-counterbalanced heavy-duty mechanism prevents vibration, allowing the unit to run smoothly with heavy, out-of-balance loads. Every other part, from the large rubber block vibration isolator feet, to the thick machined aluminum platform, is designed to last through years of nonstop use.
  • Triple counter-balanced mechanism provides optimum handling of full or unbalanced loads, regardless of flask placement, and completely eliminates “walking”
  • Microprocessor controller displays run and setpoints simultaneously on a 20-character backlit display
  • Easy programming with icon-based keys and alphanumeric messages
  • Oversized adjustable, suction-cup style, rubber feet minimize vibration, provide maximum stability, even with heavy or unbalanced loads
  • Heavy-duty, triple counter-balanced eccentric drive allows extended speed ranges
  • Low-heat 24V DC motor with moisture-resistant electronics provides continuous maintenance-free shaker operation on the benchtop or in a chamber
  • Heavy-duty, permanently lubricated bearings ensure long life, minimizing the risk of downtime
  • Optional platforms are constructed of 0.31 in., heavy-duty, brushed anodized aluminium for durability and easy cleaning
Promotional Package:
With Platform and Clamp Starter Kit
Certifications
CE, UL, cUL
Warranty and Services
One-year parts, five-years labor, and lifetime on the drive mechanism
Specifications
Specifications
25 to 525rpm
24 in.
60cm
0.5A
80% RH at or below 31C (87.8F), decreasing linearly to 50% RH at 40C (104F)
115V
19.8cm
24.3 in.
CE, cUL, UL
115V 50/60Hz
2.54cm
18 in.
45.7cm
50/60Hz
Continuous or timed from 1 min. up to 199 hr. 59 min.
7.8 in.
Tabletop orbital shaker
61.7cm
Microprocessor