Thermo Scientific™ Microm™ Air Filtration System
Features a small footprint and three speed settings for ease of use
Manufacturer: Thermo Scientific™ 58121
 View more versions of this product
Catalog No. 581-21
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Product Type Filter
    For Use With (Application) Help to minimize exposure to fumes from common laboratory solvent
    For Use With (Equipment) Microm Air Filtration System
  • Description & Specifications
Related Products
Product Content Feedback