Learn More
Thermo Scientific™ Microplate Readers Optical Filters
Manufacturer:  Thermo Scientific™ 1425600
 View more versions of this product
Catalog No. 14-386-41
Add to cart
Description
Description
  • All filters have a 3nm bandpass centered at the specified wavelength
  • Compatible with all three Multiskan models
Specifications
Specifications
560nm