Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Thermo Scientific™ Microplate Readers Optical Filters
Manufacturer:  Thermo Scientific™ 1425700
 View more versions of this product
Catalog No. 14-386-42
Description
Description
  • All filters have a 3nm bandpass centered at the specified wavelength
  • Compatible with all three Multiskan models
Specifications
Specifications
570nm