Learn More
Thermo Scientific™ Mini Gel Incubation Trays
Incubate mini-sized protein gels and Western blot membranes for staining, washing, probing and other steps of detection procedures using these convenient plastic Thermo Scientific™ Mini Gel Incubation Trays.
Manufacturer:  Thermo Scientific™ 22843
Catalog No. PI22843
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Mini Gel Incubation Trays are a convenient way to process mini-sized protein gels and Western blot membranes for use in general immunostaining applications. The trays are reusable, optically clear, and supplied with a sealable lid.
Specifications
Specifications
Clear, semi-rigid plastic (APET, 0.6mm thick) trays (95mm x 95mm base) and snap-on lids
Incubating mini-sized protein electrophoresis gels
Mini Gel Incubation Trays
10 Each