Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalgene™ Adjustable FEP Solvent Wash Bottles with Hang Tag

Click to view available options
Capacity (Metric):
1000 mL

Includes

Polypropylene hang tags (available in several colors) let you color code reagents. Write on or affix labels to the hang tags.

Ideal for rinsing with or dispensing strong solvents, acids, organic solvents, or oxidizers/peroxides

  • FEP construction
  • PTFE membrane vents vapors to reduce pressure buildup
  • ETFE screw closure and attached FEP stem and draw tube
  • Useful over a temperature range from -105° to +150°C (-157° to +302°F).
  • Transparent
  • Autoclavable

Order Info

Tags with 16 oz. (500mL) bottles accommodate 1.5 × 2 in. labels; tags with 32 oz. (1000mL) bottles accommodate 2 × 3 in. labels.

Capacity 32 oz., 1000mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Closure Material ETFE
Color Clear
Material FEP with PTFE Membrane
Closure Color Red
Closure Size 53mm