Learn More
Thermo Scientific™ Nalgene™ Centrifuge Bottle Sealing Cap Assembly
Click to view available options
For Use With (Equipment):
1L PPCO Nalgene centrifuge bottles (3141-1006)
Includes:
Red silicone gasket
Description
Includes
Includes polypropylene screw cap and shoulder ring, Tefzel™ ETFE plug, and Viton™ fluoroelastomer O-ring to form a leakproof† seal
- For use with 1L centrifuge bottles
- Autoclavable
- Polypropylene screw closure
- For use at recommended maximum speeds (10,000 x g or greater) without leakage, even when full
Specifications
For Use With (Equipment) | 1L PPCO Nalgene centrifuge bottles (3141-1006) |
Includes | Red silicone gasket |
Material | Polypropylene |