Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Thermo Scientific™ Nalgene™ Centrifuge Bottle Sealing Cap Assembly
Seals centrifuge bottle
Manufacturer:  Thermo Scientific™ DS31320RED
 View more versions of this product
Catalog No. 03-313-109
Includes: Includes polypropylene screw cap and shoulder ring, Tefzel™ ETFE plug, and Viton™ fluoroelastomer O-ring to form a leakproof† seal
Description
Description
  • For use with 1L centrifuge bottles
  • Autoclavable
  • Polypropylene screw closure
  • For use at recommended maximum speeds (10,000 x g or greater) without leakage, even when full
Specifications
Specifications
Red silicone gasket
Polypropylene
1L PPCO Nalgene centrifuge bottles (3141-1006)