Learn More
Thermo Scientific Nalgene Large Polypropylene Waste Containers with Cover
Hold and sterilize refuse, clothing, equipment, labware and contaminated material with these large polypropylene waste containers featuring secure, friction-fit closures.
Manufacturer: Thermo Scientific 69200060
Description
Hold and sterilize refuse, clothing, equipment, labware and contaminated material with Thermo Scientific Nalgene Large Polypropylene Waste Containers with Cover. These containers feature a secure, friction-fit closure and molded-in handgrips. They meet OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens.
- Molded-in handgrips make lifting easy
- Autoclavable/Biohazard
Note:
Autoclave this container upright with the lid on.
Specifications
23 L | |
Large Waste Containers with Covers, Capacity: 6 gal. | |
28 cm | |
1/Pk., 4/Cs. | |
6 gal. | |
Polypropylene |
Meet OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogensAutoclave upright with lid on. | |
11 in. | |
46 cm | |
Autoclavable | |
18 in. | |
Waste Container |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.