Learn More
Thermo Scientific™ Nalgene™ MF75™ Series Disposable Sterilization Filter Units
Triton™-free membranes to avoid cytotoxic effects, allow excellent flowrates
Manufacturer:  Thermo Scientific™ 380-0080
 View more versions of this product
Catalog No. 09-740-10
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Alert:
Nalgene CN membranes are Triton™-free
Disclaimers:
Note: CN membranes are best used for filtration of buffers and general lab solutions. We do not recommend CN for filtration of cell culture fluids because it has high protein binding characteristics. Use PES or SFCA for cell culture media, serum, additives, etc.