Thermo Scientific™ Nalgene™ Sealing Caps for Oak Ridge Centrifuge Tubes; PP screw closure, silicone gasket
Ensure leakproof service at their maximum speeds >10,000 x g, even when full
Manufacturer: Thermo Scientific™ 7131320063
 View more versions of this product
Catalog No. 03-305-080
 N/A
N/A
This product is regulated by the federal government, and may not be ordered online.  This product cannot be ordered online, as it can only be purchased internationally. 
    Specifications
    Tip Material Silicone
    Autoclavable Yes
    Material Silicone
  • Description & Specifications
Related Products
Product Content Feedback