Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Syringe Filters - 25mm Diameter

Accommodate sample volumes of 10 to 50mL

Manufacturer:  Thermo Scientific™ 1912020

 View more versions of this product

Catalog No. 09-740-35D

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

Surfactant-free Cellulose Acetate (SFCA)

  • Low protein binding
  • Ideal for sterilization and clarification of: biological/immunological samples, cell culture media and additives, and biological fluids
  • Have no wetting agents
  • Modified acrylic housings
  • Membrane of choice for sterilization
Cellulose Acetate (CA)
  • Low protein binding
  • Ideal for sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Modified acrylic housings
  • Sterile
Nylon
  • Naturally hydrophilic, low extractables
  • Useful for sterilizing cryoprotectants such as DMSO and with aqueous/biological and nonaqueous/HPLC/GC solvent applications
  • Polypropylene housings
  • Autoclavable

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit