Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Nalgene™ Syringe Filters - 25mm Diameter
Ideal for filtration of 10 to 100mL volumes
A broad choice of membranes and pore sizes are available for sterile and non-sterile laboratory applications with aqueous and non-aqueous fluids, as well as gases. Nalgene 25mm diameter syringe filters perform as well as 30mm or 33mm diameter products† in most applications, but produce less plastic waste.
$163.00 - $1810.00
Specifications
Metric Outside Diameter 25mm
Housing Material Polypropylene
View More Specs
  • Products 32
  • Description & Specifications
Products
Catalog Number Mfr. No. For Use With Application Material Pore Size Packaging Quantity Price Quantity & Availability    
09-740-120
Thermo Scientific
728-2045
Filtration of HPLC/GC samples and solvents, sterile venting - See more at: http://www.thermoscientific.com/en/product/nalgene-25mm-syringe-filters.html#sthash.K1xXUi6C.dpuf PTFE 0.45µm 100/Pk., 500/Cs. Case of 500 for $1,810.00
Check Availability
Add to Cart
09-740-108
Thermo Scientific
723-9920
Sterilization and clarification of biological/immunological samples, cell culture fluids, cellular solutions, and biological fluids SFCA 0.2µm 125/Pk., 125/Cs. Case of 125 for $398.00
Check Availability
Add to Cart
09-740-105
Thermo Scientific
723-2520
Sterilization and clarification of biological/immunological samples, cell culture fluids, cellular solutions, and biological fluids SFCA 0.2µm 50/Pk., 50/Cs. Case of 50 for $163.00
Check Availability
Add to Cart
09-740-111
NNI
724-2045
Sterilization and clarification of biological/immunological samples, cell culture fluids, cellular solutions, and biological fluids SFCA 0.45µm 100/Pk., 500/Cs. Case of 500 for $1,550.00
Check Availability
Add to Cart
09-740-112
NNI
7242080
Filtration and clarification of buffers, aqueous solutions and reagents CA 0.8µm 100/Pk., 500/Cs. Case of 500 for $1,550.00
Check Availability
Add to Cart
09-740-114
NNI
725-2545
Sterilization and clarification of cell culture media, serum and additives PES 0.45µm 50/Pk., 50/Cs. Case of 50 for $163.00
Check Availability
Add to Cart
09-740-113
NNI
725-2520
Sterilization and clarification of cell culture media, serum and additives PES 0.2µm 50/Pk., 50/Cs. Case of 50 for $163.00
Check Availability
Add to Cart
09-740-109
Thermo Scientific
7239945
Sterilization and clarification of biological/immunological samples, cell culture fluids, cellular solutions, and biological fluids SFCA 0.45µm 125/Pk., 125/Cs. Case of 125 for $398.00
Check Availability
Add to Cart
09-740-110
NNI
7242020
Sterilization and clarification of biological/immunological samples, cell culture fluids, cellular solutions, and biological fluids SFCA 0.2µm 100/Pk., 500/Cs. Case of 500 for $1,550.00
Check Availability
Add to Cart
09-740-119
Thermo Scientific
7282020
Filtration of HPLC/GC samples and solvents, sterile venting - See more at: http://www.thermoscientific.com/en/product/nalgene-25mm-syringe-filters.html#sthash.K1xXUi6C.dpuf PTFE 0.2µm 100/Pk., 500/Cs. Case of 500 for $1,810.00
Check Availability
Add to Cart
09-740-118
Thermo Scientific
727-2045
Filtering cryoprotectants such as DMSO, aqueous biological solutions, alcoholic solutions, and nonaqueous/HPLC/GC solvent applications Nylon 0.45µm 100/Pk., 500/Cs. Case of 500 for $1,550.00
Check Availability
Add to Cart
09-740-107
Thermo Scientific
723-2580
Filtration and clarification of buffers, aqueous solutions and reagents CA 0.8µm 50/Pk., 50/Cs. Case of 50 for $163.00
Check Availability
Add to Cart
09-740-117
Thermo Scientific
727-2020
Filtering cryoprotectants such as DMSO, aqueous biological solutions, alcoholic solutions, and nonaqueous/HPLC/GC solvent applications Nylon 0.2µm 100/Pk., 500/Cs. Case of 500 for $1,550.00
Check Availability
Add to Cart
09-740-106
Thermo Scientific
723-2545
Sterilization and clarification of biological/immunological samples, cell culture fluids, cellular solutions, and biological fluids SFCA 0.45µm 50/Pk., 50/Cs. Case of 50 for $163.00
Check Availability
Add to Cart
09-740-115
Thermo Scientific
7262520
Filtering cryoprotectants such as DMSO, aqueous biological solutions, alcoholic solutions, and nonaqueous/HPLC/GC solvent applications Nylon 0.2µm 50/Pk., 50/Cs. Case of 50 for $198.00
Check Availability
Add to Cart
09-740-116
Thermo Scientific
726-2545
Filtering cryoprotectants such as DMSO, aqueous biological solutions, alcoholic solutions, and nonaqueous/HPLC/GC solvent applications Nylon 0.45µm 50/Pk., 50/Cs. Case of 50 for $198.00
Check Availability
Add to Cart
09-740-35D
NNI
191-2020
N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
09-740-35J
Thermo Scientific
195-2520
Sterilizing cryoprotectants such as DMSO and with aqueous/biological and nonaqueous/HPLC/GC solvent applications 0.2µm N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
09-740-35E
NNI
1912045
N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
09-740-37F
Thermo Scientific
190-9920
Sterilization and clarification of: biological/immunological samples, cell culture media and additives, and biological fluids 0.2µm N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
09-740-37G
Thermo Scientific
1909945
Sterilization and clarification of: biological/immunological samples, cell culture media and additives, and biological fluids 0.45µm N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
09-740-35P
Thermo Scientific
196-2020
N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
09-740-38B
Thermo Scientific
199-2045
N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
09-740-35F
NNI
191-2080
N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
09-740-35Q
Thermo Scientific
196-2045
N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
09-740-61A
NNI
1942520
N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
09-740-35B
Thermo Scientific
190-2545
Sterilization and clarification of: biological/immunological samples, cell culture media and additives, and biological fluids 0.45µm N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
09-740-61B
NNI
1942545
N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
09-740-35K
Thermo Scientific
195-2545
Sterilizing cryoprotectants such as DMSO and with aqueous/biological and nonaqueous/HPLC/GC solvent applications 0.45µm N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
09-740-35A
Thermo Scientific
190-2520
Sterilization and clarification of: biological/immunological samples, cell culture media and additives, and biological fluids 0.2µm N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
09-740-38A
Thermo Scientific
199-2020
N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
09-740-35C
Thermo Scientific
190-2580
Sterile filtration and clarification of: buffers, aqueous solutions and reagents 0.8µm N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
Descriptions & Specifications
Related Products