Learn More
Thermo Scientific™ Nalgene™ Vacuum Chambers Products for Science Education
Polycarbonate jar, with white polypropylene plate.
$177.00
Specifications
For Use With (Application) For applications using nonaggressive chemicals such as student demonstrations of vacuum procedures
Includes Transparent jar and gasketed vacuum plate with a tubing adapter that fits 0.25 in. I.D. tubing
Material (Plate) Polypropylene
Type Vacuum Chamber
View More Specs
Includes: Transparent jar and gasketed vacuum plate with a tubing adapter that fits 0.25 in. I.D. tubing.
Products 1
Catalog Number Mfr. No. Capacity (English) Color Height (English) Material Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Color Height (English) Material Price Quantity & Availability  
S413811 Products for Science Education
View Documents
Thermo Scientific™
53050609/EMD
1.25 gal. Clear 9.38 in. Polycarbonate
Each for $177.00
Case of 2 EA for $352.00
Add to cart
Description
Description
  • Withstand full vacuum without risk of implosion or shattering
  • Connect to any convenient vacuum source or mechanical pump
  • System includes transparent jar and gasketed vacuum plate with tubing adapter
Avoid direct exposure of plate to chemicals. Do not expose jar to partially halogenated hydrocarbons or their vapors.
Order Info
13 in. diameter plate has a 12.63 in. O.D. gasket and adapter for 0.25 in. I.D. tubing
Specifications
Specifications
For applications using nonaggressive chemicals such as student demonstrations of vacuum procedures
Transparent jar and gasketed vacuum plate with a tubing adapter that fits 0.25 in. I.D. tubing
Polypropylene
Vacuum Chamber