Thermo Scientific™ Nalgene™ Wide-Mouth HDPE Sterile Sample Bottle; HDPE, PP screw closure
For water and wastewater samples
$179.52 - $284.29
Specifications
Material HDPE
Color Natural
Sterile Yes
Closure Material Leakproof HDPE
Certifications Certified Sterile as well as for metals and water quality parameters; manufactured under an ISO certified quality management system.
View More Specs
  • Products 2
  • Description & Specifications
Products
Catalog Number Mfr. No. English Height English Inside Diameter English Outside Diameter Closure Size Price Quantity & Availability    
05-719-359A
Thermo Scientific
N411-0125
3.88 in. 1.13 in. 2.38 in. 38mm Case of 48 for $179.52
Check Availability
Add to Cart
05-719-359B
Thermo Scientific
N411-0250
5.19 in. 1.31 in. 1.88 in. 43mm Case of 60 for $284.29
Check Availability
Add to Cart
Descriptions & Specifications
Related Products