Learn More
Thermo Scientific™ National™ Target™ Kim-Snap™ Closures
Preassembled with septa for use with Target Snap/Crimp LoVial™ Vials
Manufacturer:  Thermo Scientific™ C401164
 View more versions of this product
Catalog No. 03-397-23
  Check Availability
Add to cart
Description
Description
  • Effective evaporation resistance—apply with simple thumb pressure
  • Replace aluminum crimp seals and end tedious crimping/de-crimping
Specifications
Specifications
White
Target snap/crimp LoVial™ vials
Closure