Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ NucleoLink™ Plates, Modules and Strips

Save time and avoid contamination risk—perform solid-phase amplification and detection within the same well

Manufacturer:  Thermo Scientific™ 248450

 View more versions of this product

Catalog No. 12-566-141

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ideal for hybridization assays.

Starter Kit (Cat. No. 249344) contains everything necessary to perform testing, including: 48 strips (clear), 1 frame, 60 pieces of Tape 8, a spacer plate and application literature. These components can also be purchased separately.

  • Use in place of time-consuming methods such as gel electrophoresis and Southern blotting
  • Usable with any “ELISA-like” detection and measurement method
  • Compatible with standard microplate readers
  • Consists of 1 × 8 strips fitted into a 96-well format frame
  • V-shaped wells with flat, readable bottoms and thin walls (0.35mm)
  • Withstand temperatures from -20° to +120°C
  • Strips are sealable with polyester Tape 8
  • Working volume: Plates and BreakApart modules - 150μl, Strips 100μl

Specifications

Specifications

1 x 8
30/Cs.
Strips
Clear
8
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit