Learn More
Thermo Scientific™ Nunc™ NucleoLink™ Plates, Modules and Strips
Save time and avoid contamination risk—perform solid-phase amplification and detection within the same well
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
12-566-141
View Documents Promotion Details
Thermo Scientific™
248450
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
12-566-140
View Documents Promotion Details
Thermo Scientific™
248262
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Ideal for hybridization assays.
Starter Kit (Cat. No. 249344) contains everything necessary to perform testing, including: 48 strips (clear), 1 frame, 60 pieces of Tape 8, a spacer plate and application literature. These components can also be purchased separately.
  • Use in place of time-consuming methods such as gel electrophoresis and Southern blotting
  • Usable with any “ELISA-like” detection and measurement method
  • Compatible with standard microplate readers
  • Consists of 1 × 8 strips fitted into a 96-well format frame
  • V-shaped wells with flat, readable bottoms and thin walls (0.35mm)
  • Withstand temperatures from -20° to +120°C
  • Strips are sealable with polyester Tape 8
  • Working volume: Plates and BreakApart modules - 150μl, Strips 100μl