Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Orion 720Aplus and 920Aplus pH/mV/ISE Benchtop Meters
Includes:
- Meter Electrode Holder Only option has meter, electrode holder, instruction manual, power xupply, cord and plug for required voltage
- Meter/Electrode option includes meter, Ross Ultra combination electrode, instruction manual, cord and plug
- Meter/Manual Only option has meter, instruction manual. Requires power xupply, cord and plug for required voltage (oeder separately)
- Meter/Adapter Only option has meter, instruction manual, power xupply, cord and plug for required voltage
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
13-641-253
|
Thermo Scientific
0720A0 |
N/A | 720Aplus benchtop Dual-Channel 110V meter w/pH electrode |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
- Direct ISE and pH readout
- Simultaneous data/temperature display
- Stability display
- Selectable resolution
- Absolute and relative mV modes
- Adjustable isopotential point
- pH autocalibration; calibrate at up to five pH/ISE points
- Auto blank correction
- Manual or automatic temperature compensation with optional probe
- Self-diagnostics and user prompts
- Choice of printout options at ready, at timed intervals, or on demand
- Memory storage for calibration and data points
- Two-way RS-232-C port
- Models 920 Aplus and 720Aplus provide dual inputs, store two sets of calibration in memory; have recorder and KF outputs and numeric keypad
- Model 920Aplus permits incremental ISE techniques; Model 920Aplus gives temperature-corrected ISE readout and electrode identification; has calibration edit function for changing one or more calibration points without doing a complete calibration
- Autobuffer recognition and 25-point datalogging including time/date of last calibration; have vacuum fluorescent display
Warranty and Services
Three years
Selectable to 0.1, 0.01 or 0.001pH.
Relative mV range: 0 to ±1999.9mV.
n = number of electrons exchanged in electrode reaction process.
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title