Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents

$67.88 - $487.14

Specifications

Concentration 10ppm
View More Specs
  • Products 14
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  

13-641-883

Available on GSA/VA Contract for Federal Government customers only. 

951211
pH Adjusting ISA for Ammonia Electrode, 475mL
Each for $121.36
Check Availability
Add to Cart

13-641-852

Available on GSA/VA Contract for Federal Government customers only. 

940011
ISA for Bromide, Cadmium, Chloride, Cupric, Iodide, Sufide, Thiocyanate Electrodes, 475mL
Each for $98.93
Check Availability
Add to Cart

13-641-770

Available on GSA/VA Contract for Federal Government customers only. 

950210
ISA for Carbon Dioxide Electrode, 475mL
Each for $116.86
Check Availability
Add to Cart

13-641-954

Available on GSA/VA Contract for Federal Government customers only. 

934610
ISA for Nitrite Electrode, 475mL
Each for $101.24
Check Availability
Add to Cart

13-641-748

Available on GSA/VA Contract for Federal Government customers only. 

841111
ISA for Sodium Electrode, 475mL
Each for $67.88
Check Availability
Add to Cart

NC9975931

Thermo Scientific
951213
Storage Solution for Ammonia Electrode, 475mL
Each for $70.68
Check Availability
Add to Cart

13-641-878

Available on GSA/VA Contract for Federal Government customers only. 

941709
Complextion Ionic Strength Adjuster (CISA) for Chloride Electrode, 2 x 1L
Each for $312.02
Check Availability
Add to Cart

13-641-854

Available on GSA/VA Contract for Federal Government customers only. 

977011
Acid Reagent for Residual Chlorine Electrode, 475mL
Each for $97.09
Check Availability
Add to Cart

13-641-823

Available on GSA/VA Contract for Federal Government customers only. 

977010
Iodide Reagent for Residual Chlorine Electrode, 5 x 50mL
Each for $112.51
Check Availability
Add to Cart

13-641-853

Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
951011
Alkaline Reagent for Cyanide Electrode, 475mL
Each for $119.74
Check Availability
Add to Cart

13-642-578

Available on GSA/VA Contract for Federal Government customers only. 

940909
TISAB II for Fluoride Electrode, 1 Gallon (3.8L)
Each for $151.86
Check Availability
Add to Cart

13-641-873

Available on GSA/VA Contract for Federal Government customers only. 

940999
TISAB II for Fluoride Electrode, 4 x 1 Gallon (4 x 3.8L)
Case of 4 for $487.14
Check Availability
Add to Cart

13-641-874

Available on GSA/VA Contract for Federal Government customers only. 

940911
TISAB III Concentrate for Fluoride Electrode, 475mL
Each for $178.48
Check Availability
Add to Cart

13-641-802

Available on GSA/VA Contract for Federal Government customers only. 

930710
Nitrate Interference Suppressor Solution (NISS) for Nitrate Electrode, 475mL
Each for $99.58
Check Availability
Add to Cart
Descriptions & Specifications

Related Products