Learn More
Thermo Scientific™ Orion™ Replacement Probes
Specifications
For Use With (Application) DO
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
13-302-24
View Documents
Thermo Scientific™
083025A
N/A For Models 862A, 835A, 830A; 10m (32-3/4 ft.) cable This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • With automatic temperature compensation and convenient screw-on membrane caps
  • Rugged, for field or laboratory use
Specifications
Specifications
DO