Learn More
Thermo Scientific™ Orion™ Test Kits for AQUAfast™ IV Colorimeters
Manufacturer:  Thermo Scientific™ AC4006
 View more versions of this product
Catalog No. 13-642-164
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
EPA
0 to 0.4mg/L
30.0
AQUAfast™ IV Colorimeter Reagent Kit
AQUAfast™ IV colorimeter
Pyridine-Barbituric Acid