Thermo Scientific™ Orion™ Test Kits for AQUAfast™ IV Colorimeters
Manufacturer: Thermo Scientific™ AC4096
 View all options for this product
Catalog No. 13-642-177
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    • Kits offer premeasured reagents in bar-coded Auto-Test™ cuvets
    • AQUAfast IV Colorimeter has an Auto-ID™ function that reads the bar code to identify the species of interest automatically, then selects proper method, wavelength and time
    • Just read results on built-in display
    • Cuvet is opened only when measurement is taken, so reagent quality is not affected
  • Description & Specifications
Related Products
Product Content Feedback