Learn More
Thermo Scientific™ Orion™ Turbo2™ Balance Interface Cables
Products 3
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
13-642-732
View Documents
Thermo Scientific™
319035040
N/A Cable Kf To Mettler Ae Balance This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
13-642-735
View Documents
Thermo Scientific™
319035043
N/A Cable Kf To Mettler At Balance This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
13-642-730
View Documents
Thermo Scientific™
319035038
N/A Cable F/balances This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Enable connection of Turbo2, AF8, AF7 and AF7LC titrators to balances listed. The Cahn™/Thermo Orion CO-10 balance requires 13-642-730 Cable.