Learn More
Thermo Scientific™ Owl™ ADJ1 Adjustable System Accessories and Replacement Parts Available on GSA/VA Contract for Federal Government customers only.
Extremely compact at only 16cm wide
Manufacturer:  Thermo Scientific™ ADJ125G
 View more versions of this product
Catalog No. 09-528-169G
Add to cart
Description
Description
  • The height of the upper buffer chamber (UBC) may be adjusted by sliding the UBC up or down two support poles allowing gels to vary in height from 14 cm to 40 cm.
  • The notched or offset glass plate of the gel sandwich presses against a silicone gasket in the UBC for a leakproof seal.
  • Plates are clamped to the UBC with either binder clamps or heavy duty pony clamps.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
15.7 in.
40cm
6.2 in.
Blank Glass Plate
Glass
16cm
16 x 25cm
0.18 in.
0.48cm
16cm
Owl™ ADJ1 adjustable electrophoresis system
6.2 in.
15.7 x 6.2 in. (40 x 16cm)