Learn More
Thermo Scientific PackEye Radiation Detection Backpack
For rapid detection and location of gamma emitting radioactive sources in large areas, providing survey teams with a tool to address orphaned sources, radiological contamination, and maliciously introduced sources
Manufacturer: Thermo Scientific 4255053
Includes:
- Two He-3 counting tubes
- Intelligent amplifier FHT 681
- Connection cables to PC or notebook (serial RS232 or USB)
- Battery pack
- Earphone socket
- Optional single earphone
- Indication unit
- Voltage divider
- Photomultiplier
- NBR detector
- Set of international power socket plugs for charger
- Battery charger with inter-changeable plugs
- Connector to battery pack
- Car adapter as alternative power supply for the battery charger
Requires :
- RadEye Personal Radiation Detector (PRD)
- Data display
- Alarm indication unit
- PDA with Bluetooth communication for data storage
- Standard earphone can be used
Description
- High sensitivity plastic detector for locating radioactive sources
- Natural Background Rejection (NBR) has rapid response time and discriminates between natural (NORM) and artificial sources
- Automatic adjustment to background variations
- No false alarms on NORM and natural background changes
- High neutron detection sensitivity
- User friendly, simple LED status indication
- Unchallenged light weight of 6kg
- Low power consumption - operation time 60 to 70 hr.
- Can be tied into the ViewPoint Enterprise platform, allowing for a customizable remote monitoring solution which will provide real-time command and control data
- Display recognizes excess of high energy radiation
- Acoustic or LED bar indicator alarm; can provide a stealth mode using standard ear-phone
- Remote indication of alarm via wireless vibrator modules available
Homeland security forces, civil defense units, military, police, hazmat, security organizations, counter terrorism
Specifications
PackEye with 1Bar He-3 Ndetector |