Learn More
Thermo Scientific™ PackEye Radiation Detection Backpack
For rapid detection and location of gamma emitting radioactive sources in large areas, providing survey teams with a tool to address orphaned sources, radiological contamination, and maliciously introduced sources
Includes:
- Two He-3 counting tubes
- Intelligent amplifier FHT 681
- Connection cables to PC or notebook (serial RS232 or USB)
- Battery pack
- Earphone socket
- Optional single earphone
- Indication unit
- Voltage divider
- Photomultiplier
- NBR detector
- Set of international power socket plugs for charger
- Battery charger with inter-changeable plugs
- Connector to battery pack
- Car adapter as alternative power supply for the battery charger
Requires:
- RadEye Personal Radiation Detector (PRD)
- Data display
- Alarm indication unit
- PDA with Bluetooth communication for data storage
- Standard earphone can be used
Catalog Number | Mfr. No. | Item Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Item Description | Price | Quantity & Availability | |||||
19-175-638
|
Thermo Scientific™
4255053 |
PackEye with 1Bar He-3 Ndetector | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
19-175-639
|
Thermo Scientific™
425505091 |
PackEye PDA | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- High sensitivity plastic detector for locating radioactive sources
- Natural Background Rejection (NBR) has rapid response time and discriminates between natural (NORM) and artificial sources
- Automatic adjustment to background variations
- No false alarms on NORM and natural background changes
- High neutron detection sensitivity
- User friendly, simple LED status indication
- Unchallenged light weight of 6kg
- Low power consumption - operation time 60 to 70 hr.
- Can be tied into the ViewPoint Enterprise platform, allowing for a customizable remote monitoring solution which will provide real-time command and control data
- Display recognizes excess of high energy radiation
- Acoustic or LED bar indicator alarm; can provide a stealth mode using standard ear-phone
- Remote indication of alarm via wireless vibrator modules available
Homeland security forces, civil defense units, military, police, hazmat, security organizations, counter terrorism
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.