Thermo Scientific™ personal DataRAM™ pDR-1000AN Monitor
Palm-sized unit measures mass concentrations of dust, smoke, mists, and fumes in real time and sounds an audible alarm whenever user-defined level is exceeded
Utilizing single beam nephelometry, the pDR-1000AN incorporates light-scattering to produce optimal particle response for high correlation with standard gravimetric measurements. With datalogging enabled, the pDR-1000AN automatically tags and time stamps the collected data to be stored for subsequent retrieval, printing or computer graphing.
$5379.09
Includes: Universal power supply, PC communications software, zeroing kit, belt clip kit, carrying case, signal output cables, and instruction manual.
  • Products 1
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  
19165920
THERMO SCI SAFETY
PDR-1000AN
Personal DataRAM pDR-1000AN
Each for $5,379.09
Check Availability
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Descriptions & Specifications
Related Products