Available on GSA/VA Contract for Federal Government customers only. Thermo Scientific™ PHARM-TROL Count Control Particle Standards
For USP <788> and <789> verification
PHARM-TROL contains NIST traceable size standards with a measured particle count. The product was developed for manufactures of parenteral drugs and ophthalmic solutions seeking interim verification of USP <788> and <789> verification (Particulate Matter in Injections and Particulate Matter in Ophthalmic Solutions). The particle diameters are traceable to the Standard Meter through the National Institute of Standards and Technology (NIST).
PHARM-TROL is prepared with the same exacting procedures as the USP Count Reference Standard and provides process control data for particle counting consistency and verification of size calibration.
$314.40 - $800.40
Specifications
Metric Outside Diameter 15micro m
Material Polystyrene
Additive Contains trace amount of surfactant
Product Stability Product stable for a minimum of 12 months
Metric Density 1.05g/cm3
View More Specs
  • Products 2
  • Description & Specifications
Products
Catalog Number Mfr. No. Quantity Price Quantity & Availability    
09-980-235
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
CS-PK
6 x 25mL Bottles Pack of 6 for $314.40
Check Availability
Add to Cart
09-980-236
Available on GSA/VA Contract for Federal Government customers only. 
Thermo Scientific
CS-BX
20 x 25mL Bottles Pack of 20 for $800.40
Check Availability
Add to Cart
Descriptions & Specifications
Related Products