Thermo Scientific™ Pierce™ BCA™ Protein Assay
Faster and easier than the classic Lowry method
Used in more labs than any other detergent-compatible protein assay, Pierce BCA Reagents provide accurate determination of protein concentration with most sample types encountered in protein research. The Pierce BCA Assay can be used to assess yields in whole cell lysates and affinity-column fractions, as well as to monitor protein contamination in industrial applications. Compared to most dye-binding methods, the BCA Assay is affected much less by protein compositional differences, providing greater protein-to-protein uniformity.
$46.43 - $459.99
Includes: Kits contain BCA Reagent A, BCA Reagent B, and 10 ampules of Albumin Standard (2mg/mL)
  • Products 7
  • Description & Specifications
Products
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability    
PI-23221
Thermo Scientific
23221
Reagent A 250mL Each for $61.19
Check Availability
Add to Cart
PI-23222
Thermo Scientific
23222
Reagent A 3.75L Each for $459.99
Check Availability
Add to Cart
PI-23223
Thermo Scientific
23223
Reagent A 1L Pack for $135.76
Check Availability
Add to Cart
PI-23224
Thermo Scientific
23224
Reagent B 25mL Pack for $46.43
Check Availability
Add to Cart
PI-23227
Thermo Scientific
23227
BCA Protein Assay Kit 500mL kit Each for $119.84
Check Availability
Add to Cart
PI-23228
Thermo Scientific
23228
Reagent A 500mL Each for $89.80
Check Availability
Add to Cart
PI-23225
Thermo Scientific
23225
Protein assay kit 1L kit Pack for $186.01
Check Availability
Add to Cart
Descriptions & Specifications
Related Products