Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD23, Recombinant Protein, Invitrogen™

Recombinant Protein

Manufacturer:  Invitrogen RP38966

Catalog No. PIRP38966

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CD23/FCER2 Recombinant Protein, RP-38966, from Invitrogen.

CD23 is a 45kDa glycoprotein, which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukemia and some cases of centroblastic/centrocytic lymphoma. This product is a GST-tagged recombinant protein corresponding to human CD23, aa 48-248. On SDS-PAGE commassie blue stained gel, the purified recombinant protein shows a band at 48kDa.
Specifications

Specifications

P06734
Control
Control
0.05M tris HCl with 10mM L-glutathione and no preservative; pH 7.5
Human CD23
purified
E. coli
-20°C or -80°C if preferred
Human CD23
Human
Hu
Recombinant
Recombinant
0.1mg/mL
Liquid
2208
7.5
10μg
N/A
RUO
FCER2
Unlabeled
E. coli
Safety and Handling

Safety and Handling

ShelfLife : 12 Months

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Store at -20°C. Avoid freeze/thaw cycles.