Thermo Scientific™ Pierce™ GST Protein Interaction Pull-Down Kit
Purify protein interaction complexes with user-supplied GST-tagged fusion protein
The GST Protein Interaction Pull-Down Kit contains the necessary components to capture and purify proteins that interact with recombinant proteins containing a glutathione S-transferase (GST) fusion tag. You provide the tagged fusion protein (bait) and cells expressing the putative protein interaction target (prey), and the pull-down kit supplies everything else: cell lysis buffer, microcentrifuge spin columns, glutathione affinity resin (agarose beads), optimized binding and elution buffers, and a detailed protocol. This kit is designed to teach the method to first-time users and to increase ease-of-use, convenience and reproducibility for experienced researchers.
$378.75
Includes: Glutathione agarose resin, lysis buffer, TBS pack, glutathione for elution, microcentrifuge spin columns and collection tubes
  • Products 1
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  
PI-21516
Thermo Scientific
21516
GST Pull-Down Kit; 25-rxn kit
Each for $378.75
Check Availability
Add to Cart
Descriptions & Specifications
Related Products