Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-15, Recombinant Protein, Invitrogen™
Recombinant Protein
Supplier: Thermo Scientific RP75623
Description
IL-15 (human) Recombinant Protein, RP-75623, from Invitrogen.
RP-75623 contains purified recombinant human IL-15 protein with an N-terminal hexahistidine tag. RP-75623 may be used as a positive control in many immunological assays.Specifications
P40933 | |
Control, Western Blot | |
3600 | |
8 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Human IL-15 | |
Human | |
Hu | |
Recombinant | |
Recombinant |
0.25mg/mL | |
TBS with 50% glycerol and no preservative; pH 8 | |
Human IL-15 | |
10μg | |
RUO | |
IL15 | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
Safety and Handling
ShelfLife : 12 Months
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction