Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-14, Propeptide Region, Invitrogen™
Applications: ELISA, IHC, IP, WB
Supplier: Thermo Scientific OPA108142
Description
Host Species: Rabbit
Cross Reactivity: Hu
Immunogen: Synthetic peptide based on the propeptide region of human MMP-14.
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
MMP-14 | |
1mg/mL | |
Rabbit | |
Affinity Purified | |
Primary |
Polyclonal | |
Unconjugated | |
Synthetic peptide based on the propeptide region of human MMP-14. | |
100 μg | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title