Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Human MMP-24 Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP77536
Description
MMP24 Recombinant Protein, RP-77536, from Invitrogen.
RP-77536 is a purified recombinant MMP-24 (catalytic domain) protein purified from E. coli periplasm. MT5-MMP catalytic domain is produced by activation of a recombinant soluble proform of MT5-MMP. Activation generates two molecular species. One species begins at the N-terminus with Tyr156 of full-length MT5-MMP, while the other species is two amino acid residues shorter and starts with Leu158. Both species extend to Ser351 of MT5-MMP and they contain an additonal His-tag sequence at the C-terminal. The specific activity of MT5-MMP catalytic domain is greater than 120 mU/mg. 1 U is the activity that hydrolyzes 1μmol peptide (7-methoxycoumarin-4-yl) acetyl-Pro-Leu-Gly-Leu-(3-[2,4-dinitrophenyl]-L-2,3- diamino-propionyl)-Ala-Arg-NH2 (Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg) within 1 min. The catalytic domain of MT5-MMP is inhibited by tissue inhibitor of matrix metalloproteinase-2 (TIMP-2) and by chelators of divalent cations like EDTA or o-phenanthroline.Specifications
Q9Y5R2 | |
TBS with 5mM CaCl2 and no preservative; pH 7.5 | |
Human MMP24 | |
10μg | |
-80°C, Avoid Freeze/Thaw Cycles | |
MT5-MMP | |
MMP24 | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
Control | |
10893 | |
7.5 | |
E. coli | |
RUO | |
Human MMP24 | |
Human | |
Hu | |
Recombinant | |
Recombinant |
Safety and Handling
ShelfLife : 12 Months
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
in vitro research only
Spot an opportunity for improvement?
Provide Content Correction