Learn More
Human FKBP12 (aa 1-13), Synthetic Peptide, Invitrogen™
Synthetic Peptide
Manufacturer:  Invitrogen PEP012
Catalog No. PEP012
Add to cart
Description
Description
Human FKBP12 (residues1-13) Synthetic Peptide, PEP-012, from Invitrogen.
PEP-012 is a 14 amino acid (amino acids 1-13) synthetic peptide whose sequence is from the amino end of the human FKBP12 protein. The sequence of this peptide is (amino to carboxy terminus): G1-V-Q-V-E-T-I-S-P-G-D-G-R13-C (The C-terminal cysteine has been added to facilitate conjugation and is not a part of the native human FKBP12 sequence)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and FKBP12, catalog # PA1-026A. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.
Specifications
Specifications
Synthetic Peptide
purified
50μg
Control
peptide with no preservative
G1 - V - Q - V - E - T - I - S - P - G - D - G - R13 - C
RUO
No preservative
FKBP1A
Control
2280
Human
0.5 mg/mL
Lyophilized
Y
Human
-20° C, Avoid Freeze/Thaw Cycles
Human FKBP12 (aa 1-13)
P62942
Peptide