Learn More
Mouse Furin Convertase (aa 12-27), Synthetic Peptide, Invitrogen™
Synthetic Peptide
Manufacturer:  Invitrogen PEP010
Catalog No. PEP010
Add to cart
Description
Description
Mouse Ferin Convertase (residues 12-27) Synthetic Peptide, PEP-010, from Invitrogen.
PEP-010 is a 14 amino acid (amino acids 780-793) synthetic peptide whose sequence is from the carboxyl end of the mouse furin convertase protein. The sequence of this peptide is (amino to carboxy terminus): R-G-E-R-T-A-F-I-K-D-Q-S-A-LThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and furin convertase, catalog # PA1-062 and PA1.1-062. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.
Specifications
Specifications
Synthetic Peptide
Mouse
0.5 mg/mL
Lyophilized
Y
Mouse
-20° C, Avoid Freeze/Thaw Cycles
Mouse Furin Convertase (aa 12-27)
Peptide
purified
50μg
Control
peptide with no preservative
R - G - E - R - T - A - F - I - K - D - Q - S - A - L
RUO
No preservative
Control