Learn More
Rat MINT3 (aa 1-19), Synthetic Peptide, Invitrogen™
Synthetic Peptide
Manufacturer:  Invitrogen PEP135
Catalog No. PEP135
Add to cart
Description
Description
Rat Mint3 (residues 1-19) Synthetic Peptide, PEP-135, from Invitrogen.
PEP-135 is a 19 amino acid synthetic peptide whose sequence corresponds to amino acid residues 1-19 from rat Mint3 protein. The sequence of this peptide is (amino to carboxy terminus): M (1)-E-F-L-P-E-P-Q-H-P-P-G-P-P-T-M-D-L- E (19).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and rat Mint3 protein, catalog # PA1-072. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (~70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
Specifications
Specifications
Rat
83611
Rat
0.5 mg/mL
Lyophilized
Y
RUO
No preservative
Apba3
Control
Synthetic Peptide
purified
50μg
Control
peptide with no preservative
M(1) E F L P E P Q H P P G P P T M D L E(19)
-20° C, Avoid Freeze/Thaw Cycles
Rat MINT3 (aa 1-19)
O70248
Peptide