Learn More
Mouse NCoR1 (aa 2437-2453), Synthetic Peptide, Invitrogen™
Synthetic Peptide
Manufacturer:  Invitrogen PEP061
Catalog No. PEP061
Add to cart
Description
Description
Mouse N-CoR (residues 2437-2453) Synthetic Peptide, PEP-061, from Invitrogen.
PEP-061 is an 18 amino acid synthetic peptide whose sequence corresponds to residues 2437-2453 of the mouse N-CoR protein with an N-terminal cysteine added for immunogen preparation. The sequence of this peptide is (amino to carboxy terminus): C-P2437-A-P-L-L-S-A-Q-Y-E-T-L-S-D-S-D-D2453This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and N-CoR, catalog # PA1-844A. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (~70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
Specifications
Specifications
Synthetic Peptide
20185
Mouse
0.5 mg/mL
Lyophilized
Y
RUO
No preservative
Ncor1
Control
Mouse
purified
50μg
Control
peptide with no preservative
C - P2437 - A - P - L - L - S - A - Q - Y - E - T - L - S - D - S - D - D2453
-20° C, Avoid Freeze/Thaw Cycles
Mouse NCoR1 (aa 2437-2453)
Q60974
Peptide