Learn More
Human 20S Proteasome Y (aa 41-58), Synthetic Peptide, Invitrogen™
Synthetic Peptide
Manufacturer:  Invitrogen PEP173
Catalog No. PEP173
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Human 20S Proteasome Y (residues 41-58) Synthetic Peptide, PEP-173, from Invitrogen.
PEP-173 is an 18 amino acid synthetic peptide whose sequence is derived from human proteasome 20S Y and amino acid residues 74-91 from frog protein. The sequence of this peptide is (amino to carboxy terminus): C (41)-R-S-G-S-A-A-D-T-Q-A-V /I-A-D-A-V-T-Y (58)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human proteasome 20S Y, catalog # PA1-978. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (~70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
Specifications
Specifications
Synthetic Peptide
5694
Human
0.5 mg/mL
Lyophilized
Y
RUO
No preservative
PSMB6
Control
Human
purified
50μg
Control
peptide with no preservative
C(41)-R-S-G-S-A-A-D-T-Q-A-V /I-A-D-A-V-T-Y(58)
-20° C, Avoid Freeze/Thaw Cycles
Human 20S Proteasome Y (aa 41-58)
P28072
Peptide