Learn More
Mouse RARA (aa 444-462), Synthetic Peptide, Invitrogen™
Synthetic Peptide
Manufacturer:  Invitrogen PEP004
Catalog No. PEP004
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Mouse Retinoic Acid (residues 444-462) Synthetic Peptide, PEP-004, from Invitrogen.
PEP-004 is a synthetic peptide whose sequence is from the C-terminal end of mouse retinoic acid receptor alpha (RAR alpha). The 19 amino acid sequence extends from amino acids 444 to 462 of RAR alpha. The sequence of PEP-004 is (amino to carboxy terminus): C-S-P-S-L-S-P-S-S-H-R-S-S-P-A-T-Q-S-PThis peptide may be used for neutralization and control experiments with the monoclonal and polyclonal antibodies that react with this product and human and mouse RAR*, catalog # MA1-810A and PA1-810A. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.
Specifications
Specifications
Synthetic Peptide
purified
50μg
Control
peptide with no preservative
C - S - P - S - L - S - P - S - S - H - R - S - S - P - A - T - Q - S - P
RUO
No preservative
Rara
Control
19401
Mouse
0.5 mg/mL
Lyophilized
Y
Mouse
-20° C, Avoid Freeze/Thaw Cycles
Mouse RARA (aa 444-462)
P11416
Peptide