Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GRASP55 (aa 419-433), Synthetic Peptide, Invitrogen™
Click to view available options
Quantity:
50 μg
Description
Mouse GRASP55 (residues 419-433) Synthetic Peptide, PEP-295, from Invitrogen.
PEP-295 is a 15 amino acid synthetic peptide whose sequence corresponds to residues 419-433 of mouse GRASP55. The sequence of this peptide is (amino to carboxyl terminus): P (419) A S K V P T T V E D R V S D (433).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, GRASP55, catalog # PA1-076.

Specifications
Specifications
Gene ID (Entrez) | 70231 |
Species | Mouse |
Purification Method | HPLC |
Accession Number | Q99JX3 |
Common Name | Mouse GRASP55 (aa 419-433) |
Content And Storage | -20°C. Avoid freeze-thaw cycles. |
Description | Synthetic Peptide |
Format | Liquid |
Formulation | peptide with no preservative |
For Use With (Application) | Control |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction