Thermo Scientific™ Pierce™ Normal Serum
Non-immune sera from goat, human, mouse, rabbit and other species
Normal serum that is derived from the same or related species of animal as a tissue sample or from the host species of specific antibodies used in immunodetection method provides several properties that improve assay performance for immunohistochemistry. Normal serum provides a sufficient quantity of endogenous proteins to saturate and block nonspecific binding sites on fixed samples or assay substrates. When used as an antibody diluent, non-immune serum also supplies a native chemical environment for antibodies to function properly without adding foreign protein components.
$37.99 - $104.45
Specifications
Formulation Lyophilized powder from delipidated and PBS-dialyzed serum
View More Specs
  • Products 10
  • Description & Specifications
Products
Catalog Number Mfr. No. Quantity Price Quantity & Availability    
PI-31872
Thermo Scientific
31872
2mL Pack for $37.99
Check Availability
Add to Cart
PI-31873
Thermo Scientific
31873
10mL Pack for $104.45
Check Availability
Add to Cart
PI-31874
Thermo Scientific
31874
2mL Pack for $37.99
Check Availability
Add to Cart
PI-31876
Thermo Scientific
31876
2mL Pack for $37.99
Check Availability
Add to Cart
PI-31880
Thermo Scientific
31880
2mL Pack for $48.54
Check Availability
Add to Cart
PI-31881
Thermo Scientific
31881
5mL Pack for $70.69
Check Availability
Add to Cart
PI-31883
Thermo Scientific
31883
5mL Pack for $70.69
Check Availability
Add to Cart
PI-31884
Thermo Scientific
31884
2mL Pack for $37.99
Check Availability
Add to Cart
PI-31888
Thermo Scientific
31888
2mL Pack for $37.99
Check Availability
Add to Cart
PI-31890
Thermo Scientific
31890
2mL Each for $37.99
Check Availability
Add to Cart
Descriptions & Specifications
Related Products