Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Premium Grade BS3

Catalog No. PIPG82084
Click to view available options
Quantity:
1 g

Crosslink proteins and other biomolecules in applications where product integrity and risk minimization are paramount with Thermo Scientific™ Pierce™ Premium Grade BS3, a high-quality formulation of this popular amine-reactive crosslinker.

Thermo Scientific Pierce Premium Grade BS3 is our highest quality formulation of this popular amine-reactive crosslinker, specially characterized for applications where product integrity and risk minimization are paramount. BS3 is a noncleavable, membrane-impermeable, water-soluble crosslinking agent for amines.

  • High quality - identity and purity confirmed by multiple tests, including quantitative NMR
  • Product integrity - enhanced level of testing and characterization compared to standard grade
  • Lot retention - ample supply of past lots retained to help ensure future process testing
  • Change management - Change Control Notification (CCN) service
  • Consistent manufacture - batch-specific manufacturing documentation review
Compared to the standard grade product, Premium Grade BS3 provides more clearly defined quality and product support by including increased analytical testing and product characterization, greater batch-specific information and quality assurance review, extensive lot sample retention, and change control notification.
BS3 is bis(sulfosuccinimidyl)suberate, an amine-to-amine crosslinker that is homobifunctional, water-soluble, non-cleavable, and membrane impermeable. BS3 contains an amine-reactive N-hydroxysulfosuccinimide (NHS) ester at each end of an 8-carbon spacer arm. NHS esters react with primary amines at pH 7-9 to form stable amide bonds, along with release of the N-hydroxysulfosuccinimide leaving group. Proteins, including antibodies, generally have several primary amines in the side chain of lysine (K) residues and the N-terminus of each polypeptide that are available as targets for NHS-ester crosslinking reagents.
Applications:
  • Crosslink cell-surface proteins prior to cell lysis and immunoprecipitation
  • Identify receptor-ligand interactions by chemical crosslinking
  • “Fix” protein interactions to allow identification of weak or transient protein interactions
  • Identify near-neighbor protein interactions
  • Crosslink proteins to create bioconjugates via single-step reactions
  • Immobilize proteins onto amine-coated surfaces

TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Amine
Cleavable No
Molecular Weight (g/mol) 572.43
PEGylated No
Spacer Arm Length 11.4 Å
Content And Storage Upon receipt store with desiccant at 4°C.
Sufficient for 1.74 L of typical 1X (1 mM) reaction solution or for ∼500 uses at 2 mg reagent per use.
Cell Permeability No
Shipping Condition Ambient
Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Sulfo-NHS Ester
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 1 g
Solubility Water
Format Standard, Single-use, Premium-grade
Water Soluble Yes
Product Type BS3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.