Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Premium Grade BS3

Crosslink proteins and other biomolecules in applications where product integrity and risk minimization are paramount with Thermo Scientific™ Pierce™ Premium Grade BS3, a high-quality formulation of this popular amine-reactive crosslinker.

Specifications

Molecular Weight (g/mol) 572.43
Solubility Water
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPG82084
View Documents
Thermo Scientific™
PG82084
1 g Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View alternate product
N/A
PIPG82083
View Documents
Thermo Scientific™
PG82083
100 mg Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

Thermo Scientific Pierce Premium Grade BS3 is our highest quality formulation of this popular amine-reactive crosslinker, specially characterized for applications where product integrity and risk minimization are paramount. BS3 is a noncleavable, membrane-impermeable, water-soluble crosslinking agent for amines.

  • High quality - identity and purity confirmed by multiple tests, including quantitative NMR
  • Product integrity - enhanced level of testing and characterization compared to standard grade
  • Lot retention - ample supply of past lots retained to help ensure future process testing
  • Change management - Change Control Notification (CCN) service
  • Consistent manufacture - batch-specific manufacturing documentation review
Compared to the standard grade product, Premium Grade BS3 provides more clearly defined quality and product support by including increased analytical testing and product characterization, greater batch-specific information and quality assurance review, extensive lot sample retention, and change control notification.
BS3 is bis(sulfosuccinimidyl)suberate, an amine-to-amine crosslinker that is homobifunctional, water-soluble, non-cleavable, and membrane impermeable. BS3 contains an amine-reactive N-hydroxysulfosuccinimide (NHS) ester at each end of an 8-carbon spacer arm. NHS esters react with primary amines at pH 7-9 to form stable amide bonds, along with release of the N-hydroxysulfosuccinimide leaving group. Proteins, including antibodies, generally have several primary amines in the side chain of lysine (K) residues and the N-terminus of each polypeptide that are available as targets for NHS-ester crosslinking reagents.
Applications:
  • Crosslink cell-surface proteins prior to cell lysis and immunoprecipitation
  • Identify receptor-ligand interactions by chemical crosslinking
  • “Fix” protein interactions to allow identification of weak or transient protein interactions
  • Identify near-neighbor protein interactions
  • Crosslink proteins to create bioconjugates via single-step reactions
  • Immobilize proteins onto amine-coated surfaces

Specifications

Specifications

572.43
Water
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit