Learn More
Thermo Scientific Pierce STAT6 In-Cell ELISA Kit
Measure total and phosphorylated signal transducers and activators of transcription 6 using antibodies detected by fluorescence or TMB substrate in whole-cell plate assays.
Manufacturer: Thermo Scientific 62208
Description
Thermo Scientific Pierce STAT6 In-Cell ELISA Kits are colorimetric and fluorescent microplate assays optimized to measure total and phosphorylated signal transducers and activators of transciption 6 (phospho-STAT6) in whole cells.
Pierce STAT6 In-Cell ELISA Kits enable the relative quantitation of total and phosphorylated signal transducers and activators of transciption 6 (STAT6) in whole cells using a 96- or 384-well microplate format. The complete kits include a mouse monoclonal anti-STAT6 antibody, rabbit monoclonal anti-phospho-(Tyr641) STAT6 antibody, detection reagents and all buffers required for either colorimetric or near infrared detection.The Colorimetric In-cell ELISA Kit facilitates comparison of multiple target protein levels from different wells using target-specific antibodies, HRP-conjugated detection reagents and TMB substrate. Results across wells are then normalized to cell number based on whole-cell staining with Janus Green.
The Near Infrared In-Cell ELISA Kit allows simultaneous detection of two different target proteins within the same well, eliminating variability caused by differences in cell plating. With the near IR system, protein levels are determined by using target-specific primary antibodies and DyLight Fluor-Conjugated Secondary Antibodies. The near IR detection method eliminates issues with background fluorescence, providing a high signal-to-noise ratio and making these fluorescent multiplex experiments both sensitive and robust.
Highlights:
In-cell ELISA target-specific detection directly from plated whole cells; no cell lysis, protein extraction or special sample preparation required
Quantitative and specific system provides accurate measurement of relative protein expression levels in treated vs. nontreated cells
Better than Western blotting In-cell ELISA directly yields quantitative results and provides higher throughput than Western blotting techniques
Less expensive costs less (one well vs. one lane) than Western blotting methods of protein expression and activation analysis
Multiplex-capable replicate wells or different fluors enable multiple ELISA targets to be measured relative to the primary targets supplied with the kit
Two kit formats choose the colorimetric kit for use with standard microplate readers; choose the near infrared fluorescent detection kit for use with IR imaging systems
Includes:
Anti-Phospho STAT6 (Y641) Antibody, Anti-STAT6 Antibody, secondary antibodies, buffers, detergents and plate seals
Specifications
Pierce STAT6 Colorimetric In-Cell ELISA Kit | |
20X Tris Buffered Saline, 50mL | |
96 Test Kit |
Multi-component kit with HRP-conjugate | |
96 microplate wells of overnight-cultured cells |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.